JS-7.5 Swinging Bucket Rotor, Beckman Coulter®
Supplier: Beckman Coulter
336380
BK336380EA 16112.96 USD
BK336380
JS-7.5 Swinging Bucket Rotor, Beckman Coulter®
Centrifuges Rotors
Four-place, aluminum swinging bucket rotors are for use in Avanti™ J-26 XP and XPIBK393124 series and J6-MIBK360291 centrifuges.
  • Applications: Initial Processing of Cells and Removal of Cell Debris from Culture Media
  • Space for Four 250 mL
  • Material: Aluminum
  • Four-Place Rotors
They are ideal for initial processing of cells and removal of cell debris from culture media.
The rotors accommodate four 250 mL tubes.