JA-21 Fixed-Angle Rotor, Beckman Coulter®
Supplier: Beckman Coulter
334845
BK334845EA 8878.47 USD
BK334845
JA-21 Fixed-Angle Rotor, Beckman Coulter®
Centrifuges Rotors
This aluminum, 18-place, and fixed-angle rotor is for use in Avanti™ J-26 XP and XPIBK393124 series and J6-MIBK360291 and J-EBK369001 centrifuges.
  • Application: Separation of Many Samples in Small Volumes
  • Material: Aluminum
  • Eighteen-Place Router
  • Fixed-Angle
  • Rotor Accommodates Eighteen 10 mL Tubes
It's perfect for rapid, efficient separation of many samples in small volumes, including viruses, bacteriophages, mitochondria, and nuclei.
The rotor accommodates eighteen 10 mL tubes.