JA-20 Fixed-Angle Rotor, Beckman Coulter®
Supplier: Beckman Coulter
334831
BK334831EA 9031.41 USD
BK334831
JA-20 Fixed-Angle Rotor, Beckman Coulter®
Centrifuges Rotors
The aluminum, eight-place, and fixed-angle rotor is for use in Avanti™ J-26 XP and XPIBK393124 series and J-EBK369001 and J6-MIBK360291 centrifuges.
  • Applications: Harvesting Bacteria and Cell Membranes, Processing Tissue Homogenates, Separating Cell Particulates
  • Material: Aluminum
  • Eight-Place Router
  • Fixed-Angle
  • Router Accommodates Eight 50 mL Tubes
It's ideal for harvesting bacteria and cell membranes, processing tissue homogenates, and separating cell particulates.
This rotor accommodates eight 50 mL tubes.