Thinwall Polyallomer Tubes, Beckman Coulter
Supplier: Beckman Coulter
331372
BK331372PK 380.82 USD
BK331372
Thinwall Polyallomer Tubes, Beckman Coulter
Tubes Centrifuge Tubes
Thinwall polyallomer tubes are for use in ultracentrifuge rotors.
They have an open-top, thinwall closure type and are transparent. Made of polyallomer material.