Biomek® 96-Well Microplates, Beckman Coulter®
Supplier: Beckman Coulter
267007 267004 267006 609844
BK267007CS 188.7 USD
BK267007 BK267004 BK267006 BK609844 BK609681 BK140504
Biomek® 96-Well Microplates, Beckman Coulter®
Microplates Deep Well Microplates
Biomek® 96-Well Microplates are designed for use on all Biomek® liquid handlers. To accelerate method setup, all microplate labware properties have been predefined in Biomek® software.
  • Ideal for Large-Volume Applications
  • Acrylic Adhesive Seals to Withstand Temperatures From -50° to 130°C
  • Pre-Cut Seals
  • Specially-Formulated Polystyrene
  • Compatible with All Biomek® Liquid Handlers
The 96-Well flat microplate is manufactured from specially-formulated polystyrene and optimized for maximum protein binding with heterogeneous solid-phase assays such as ELISAs
For optimum sealing, use Biomek® Aluminum Foil Seals (BK538619) with Roller (BK538618). Biomek Cap Mat (BK267002 or BK267005) may also be used to tightly seal 96-Well Round Deep-Well plates.
It features round wells and a flat bottom with a volume capacity of 300 µL per well. Both round and square deep-well plates feature a rounded bottom to minimize dead volume. Volume capacity is 1 mL and 2 mL per well respectively, making these microplates ideal for large volume applications.