Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

X31087-2.5L
BJX31087-2.5LEA 135 USD
BJX31087-2.5L
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Hydrochloric acid

Honeywell's high-quality titration products cover the complete titration workflow and include:


  • Solutions for volumetric titration (acids, bases, or salts)Buffers available as concentrates or ready-to-useReagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
  • Masking agents
  • Indicators for volumetric and complexometric titrations

Formula: HCl
Density: 1.072 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 3,III

Order Now

Specification Test Results

Appearance (color) complying
Appearance (form) complying
APHA Max. 20
Assay 5.988 - 6.012 mol/l
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR