o-Xylene ≥98.0%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

X1040-18L
BJX1040-18LEA 911.98 USD
BJX1040-18L
o-Xylene ≥98.0%, Reagent Grade
o-Xylene

  • ≥95% for general laboratory use.


Beilstein Number: 1815558

Formula: C₈H₁₀
Boiling Pt: 7.6 %(V)
Melting Pt: –25 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 3.7 (vs air)
Identity (IR) complying
Assay (GC) Min. 98.0 %
m-, p-Xylene (GC) Max. 1.0 %
Ethyl benzene (GC) Max. 0.50 %
Toluene (GC) Max. 0.50 %
Autoignition Temperature 867 °F
Explosion Limit 7 %
Refractive Index n20/D 1.505 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR