5-(Ethylthio)-1H-tetrazole (ETT) 0.25M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
5-Ethyltio-1H-Tetrazole
BR721-4
SR721-1
SR721-1
BC721-200
BC721-56
BR721-20
BR721-4X2.5L
SR721-2.5
BJBR721-2CS
60705.6
USD
BJBR721-2
BJBR721-4
BASR721-1NAN
BJSR721-1
BJBC721-200
BJBC721-56
BJBR721-20
BJBR721-4X2.5L
BJSR721-2.5
5-(Ethylthio)-1H-tetrazole (ETT) 0.25M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazole (ETT)
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Formula:
C₃H₆N₄S MW: 130.17 g/mol Boiling Pt: 82 °C Melting Pt: -46 °C Density: 0.790 g/cm3 Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068733 CAS Number: 89797-68-2 UN: 1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...