Capping Reagent (Acetic Anhydride:Lutidine:Acetonitrile) (20:30:50), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

SR644-20
BJSR644-20EA 9916 USD
BJSR644-20
Capping Reagent (Acetic Anhydride:Lutidine:Acetonitrile) (20:30:50), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent

Reagents are specifically processed and purified to ensure the high synthesis efficiency


  • Ideal for oligonucleotide synthesis
  • Extremely low water content
  • High purity reagents
  • Low amine specifications


Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.

Density: 0.881 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
UN: 2924
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR