di-Sodium tetraborate decahydrate ≥99.5% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
S9640-2.5KG
S9640-4X2.5KG
S9640-500G
S9640-6X500G
BJS9640-2.5KGEA
183.42
USD
BJS9640-2.5KG
BJS9640-25G
BJS9640-4X2.5
BJS9640-500G
BJS9640-6X500G
di-Sodium tetraborate decahydrate ≥99.5% ACS, Fluka™
di-Sodium tetraborate decahydrate
Formula:
Na₂B₄O₇·10H₂O Melting Pt: 742 °C (anhy.) ≥ 62 °C (–H₂O of crystall.) Density: 1.7 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 ADR: 3, |
Specification Test Results
Assay | 99.5 - 105.0 % |
pH (0.01 m, 25°C) | 9.15 - 9.20 |
Water insoluble matter | Max. 0.005 % |
Calcium (Ca) | Max. 50 ppm |
Iron (Fe) | Max. 5 ppm |
Sulfate (SO₄) | Max. 50 ppm |
Heavy metals (as Pb) | Max. 0.001 % |
Chloride (Cl) | Max. 10 ppm |
Phosphate (PO₄) | Max. 10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...