Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

S5881-6X1KG S5881-500G S5881-5KG
BJS5881-1KGCS 686.45 USD
BJS5881-1KG BJS5881-500G BJS5881-5KG
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Vapor Density >1 (vs air)
Assay Min. 98.0 %
Assay of Na₂CO₃ Max. 1.0 %
Appearance of the solution complying
Turbidity of solution Max. 3.5 NTU
Vapor Pressure 3 mmHg ( 37 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR