Dichloromethane ≥99.9% stabilised, Purified Plus® ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Warning


Stabilization: Stabilised with cyclohexene

PP300-19
BJPP300-19EA 848.98 USD
BJPP300-19 BJPP300-204
Dichloromethane ≥99.9% stabilised, Purified Plus® ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Dichloromethane

For specialty organic synthesis, preparative scale liquid chromatography, glassware cleaning, and applications requiring ACS reagent grade solvent. The 54gal. drum is individually analyzed with certificate on label.


  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: CH₂Cl₂
Boiling Pt: 40 °C
Melting Pt: –95 °C
Density: 1.33 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99.9 % min.
Color (APHA) 10 max.
Free Halogens Passes test
Refractive Index (20ºC) 1.4231 - 1.4251
Residue after Evaporation 5 mg/L max.
Titrable Acid 0.3 µeq/g max.
Water by Karl Fisher Titration 0.02 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR