Acetonitrile ≥99.8%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

PP016TC19 PP016-204
BJPP016TC19EA 1499.89 USD
BJPP016TC19 BJPP016-204
Acetonitrile ≥99.8%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Acetonitrile

For specialty organic synthesis, preparative scale liquid chromatography, glassware cleaning, and applications requiring ACS reagent grade solvent. The 54-gallon drum is individually analyzed with certificate attached.


  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: H₃CCN
Boiling Pt: 82 °C(1.013 hPa)
Melting Pt: –46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99.9 % min.
Color (APHA) 10 max.
Refractive Index (20ºC) 1.3430 - 1.3450
Residue after Evaporation 5 mg/L max.
Titrable Acid 8 µeq/g max.
Titrable Base 0.6 µeq/g max.
Water by Karl Fischer Titration 0.1 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR