≥98% ACS, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

P3786-100G P3786-1KG P3786-2.5KG P3786-500G P3786-6X500G
BJP3786-100GEA 72.68 USD
BJP3786-100G BJP3786-1KG BJP3786-2.5KG BJP3786-500G BJP3786-6X500G
≥98% ACS, Fluka™
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98.0 %
Water insoluble matter Max. 0.01 %
Loss on drying (105°C) Max. 1.0 %
pH (5 %, 25 °C) 8.5 - 9.6
Iron (Fe) Max. 10 ppm
Sodium (Na) Max. 500 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 30 ppm
Total N Max. 10 ppm
Sulfate (SO₄) Max. 50 ppm
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR