Methanol ≥99.9%, B&J Brand™ ACS for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

230-1L NP230-20 230-4 NS230-20
BJ230-1LCS 588 USD
BJ230-1L BJNP230-20 BJ230-4 BJNS230-20
Methanol ≥99.9%, B&J Brand™ ACS for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Methanol

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Fluorescence (as quinine base) 0.30 ppb max.
GC Analysis 99.9 % min.
HPLC Gradient @ 220 nm 0.005 AU max.
HPLC Gradient @ 254 nm 0.001 AU max.
Refractive Index (20ºC) 1.3280 - 1.3288
Residue 1 mg/L max.
UV Cutoff 205 nm max.
Water by Karl Fischer Titration 0.05 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR