Hexane (mixture of isomers) ≥60.0% (n-hexane) ≥99.9% (by GC), B&J Brand™ for gas chromatography, for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Hexanes, Isohexane
211-4
NS211-20
BJ211-4CS
908
USD
BJ211-4
BJNS211-20
BJCS219-200
Hexane (mixture of isomers) ≥60.0% (n-hexane) ≥99.9% (by GC), B&J Brand™ for gas chromatography, for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™
Hexane (mixture of isomers)
Suitable for gas chromatography and pesticide residue analysis.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 68.7 °C Melting Pt: –95 °C Density: 0.659…0.673 g/cm³ (20 °C) Flash Pt: –26 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179311 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Specification Test Results
GC Analysis (n-Hexane) | 60 % min. |
GC Analysis (total C6 isomers) | 99.9 % min. |
Residue | 1 mg/L max. |
Water by Karl Fisher Titration | 0.01 % max. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...