Iodine 0.02 M I₂ in 70/20/10 THF/pyridine/water (v/v), BioSyn™ oxidation reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

NP666-20L
BJNP666-20LEA 1915 USD
BJNP666-20L
Iodine 0.02 M I₂ in 70/20/10 THF/pyridine/water (v/v), BioSyn™ oxidation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Iodine

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Formula: I₂
Boiling Pt: 66 °C
Melting Pt: –108.5 °C
Density: 0.935 g/cm3 (20 °C)
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR