Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

NP654-20L
BJNP654-20LEA 13677 USD
BJNP654-20L
Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Boiling Pt: 86.6 °C (main component)
Storage Temperature: Ambient
MDL Number: MFCD00001878
UN: 2924
ADR: 3,8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR