Capping Reagent 50/30/20 Acetonitrile/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

NP644-20L
BJNP644-20LEA 7980 USD
BJNP644-20L
Capping Reagent 50/30/20 Acetonitrile/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Density: 0.881 g/cm3 (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
UN: 2924

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR