Acetonitrile : Water (80:20)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

NP460-20
BJNP460-20EA 3595.21 USD
BJNP460-20
Acetonitrile : Water (80:20)%, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile : Water

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk


UNSPSC Code: 12350000

Boiling Pt: 66.1 °C
Density: 0.8298 g/cm³ (25 °C)
Flash Pt: 16 %(V) (main component)
Storage Temperature: Ambient
CAS Number: 75-05-8
UN: 1648
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR