N-Methyl-2-pyrrolidone (NMP) ≥99.5%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

304-1L 304-4 NP304-20
BJ304-1LCS 2632 USD
BJ304-1L BJ304-4 BJNP304-20 BJCS304-56
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™
N-Methyl-2-pyrrolidone (NMP)

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.0259 g/cm³ (25 °C)
Flash Pt: 9.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.5 % min.
Refractive Index (20ºC) 1.4680 - 1.4720
UV Absorbance @ 300 nm 0.5 AU max.
UV Absorbance @ 325 nm 0.1 AU max.
UV Absorbance @ 350 nm 0.03 AU max.
UV Absorbance @ 400 nm 0.01 AU max.
UV Cutoff 285 nm max.
Water by Karl Fischer Titration 0.05 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR