Methylcyclohexane 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

M37889-1L M37889-2.5L
BJM37889-1LEA 80 USD
BJM37889-1L BJM37889-2.5L BJM37889-20L
Methylcyclohexane 99%, Reagent Grade
Methylcyclohexane
Formula: C₆H₁₁CH₃
Boiling Pt: 6.7 %(V)
Melting Pt: -126 °C
Density: 0.769 g/cm³ (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 98.5 %
Vapor Pressure 37 mmHg ( 20 °C)
Refractive Index n20/D 1.422 (lit.)
Vapor Density 3.4 (vs air)
Autoignition Temperature 545 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR