Methanol absolute acetone-free

Supplier: Honeywell Research Chemicals
Danger

M1775-1GA
BJM1775-1GAEA 192.07 USD
BJM1775-1GA
Methanol absolute acetone-free
Methanol
Formula: CH₃OH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Pressure 410 mmHg ( 50 °C)
Appearance complying
Autoignition Temperature 725 °F
Refractive Index n20/D 1.329 (lit.)
Explosion Limit 36 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR