Water ACS, meets ASTM Type II specifications for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals

LP366-4P LP366-5G
BJLP366-4PCS 221.41 USD
BJLP366-4P BJLP366-5G
Water ACS, meets ASTM Type II specifications for preparative liquid chromatography, general laboratory use
Water

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9971 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Chloride (Cl) 5 µg/L max.
Electrical Conductance (25ºC) 1 µS/cm max.
Electrical Resisitivity (25ºC) 1 MOhm/cm min.
Sodium (Na) 5 µg/L max.
Total Organic Carbon 50 µg/L max.
Total Silica (SiO2) 3 µg/L max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR