N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

LP304-4P
BJLP304-4PCS 1551.66 USD
BJLP304-4P
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
N-Methyl-2-pyrrolidone (NMP)

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.0259 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99 % min.
Chloride 1 ppm max.
Color (APHA) 50 max.
Free Amines (as CH3NH2) 0.01 % max.
Water by Karl Fischer Titration 0.05 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR