tert-Butyl methyl ether ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals
Danger

LP242-4
BJLP242-4CS 905.87 USD
BJLP242-4
tert-Butyl methyl ether ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
tert-Butyl methyl ether

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


UNSPSC Code: 12191602

Formula: (CH₃)₃COCH₃
Boiling Pt: 15.1 %(V)
Melting Pt: –108.6 °C
Density: 0.74 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
UN: 2398
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99 % min.
Color (APHA) 10 max.
Peroxides (as H₂O₂) at time of packaging 1 ppm max.
Residue after Evaporation 0.001 % max.
Water by Karl Fischer Titration 0.05 % max.
SolubilityinWater(%)at20°C 4.8
SurfaceTension(dyn/cm)at20°C 19.4
VaporPressure(Torr)at20°C 240


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR