N,N-Dimethylformamide ≥99.8% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals
Danger

LP076-19
BJLP076-19EA 1389.05 USD
BJLP076-19
N,N-Dimethylformamide ≥99.8% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
N,N-Dimethylformamide

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


UNSPSC Code: 12191602

Formula: HCON(CH₃)₂
Boiling Pt: 15.2 %(V)
Melting Pt: –60.4 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99.8 % min.
Color (APHA) 10 max.
Residue after Evaporation 0.005 % max.
Titrable Acid 0.5 µeq/g max.
Titrable Base 3 µeq/g max.
Water by Karl Fischer Titration 0.15 % max.
VaporPressure(Torr)at20°C 2.7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR