Acetonitrile : water : formic acid (95:5:0.1)%, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

LC599-2.5
BJLC599-2.5CS 707 USD
BJLC599-2.5
Acetonitrile : water : formic acid (95:5:0.1)%, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Acetonitrile:Water:Formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 70 °C
Density: 0.7951 g/cm³ (20 °C)
Flash Pt: 6.67 °C
Storage Temperature: Ambient
UN: 1648
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR