Methanol with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

LC445-2.5
BJLC445-2.5CS 905.8 USD
BJLC445-2.5
Methanol with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Methanol with 0,1% (v/v) formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 64.7 °C (main component)
Melting Pt: –97.68 °C (main component)
Density: 0.792 g/cm³ (20 °C) (main component)
Flash Pt: 11 °C
Storage Temperature: Ambient
UN: 1230
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR