Methanol with 0.1% Ammonium hydroxide, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

LC2149-4
BJLC2149-4CS 1255.84 USD
BJLC2149-4
Methanol with 0.1% Ammonium hydroxide, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Methanol with 0.1% Ammonium hydroxide

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk


UNSPSC Code: 12350000

Boiling Pt: 64.4 °C
Density: 0.7865 g/cm³ (25 °C)
Flash Pt: 11.1 °C
Storage Temperature: Ambient
CAS Number: 67-56-1
UN: 1230
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR