2-Propanol ≥99.0%, meets analytical specification of USP, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

I9030-500ML
BJI9030-500MLEA 129 USD
BJI9030-500ML
2-Propanol ≥99.0%, meets analytical specification of USP, Riedel-de Haën™
2-Propanol
Formula: (CH₃)₂CHOH
Boiling Pt: 77…83 °C (1.013 hPa)
Melting Pt: –90 °C
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 2.1 (vs air)
Assay (GC) Min. 99.0 %
Density (D 25/25) 0.783 - 0.787
Refractive index (n 20/D) 1.376 - 1.378
Water (Karl Fischer) Max. 0.5 %
Non-volatile matter Max. 0.005 %
Acidity complying
Volatile impurities (GC) complying
Acetone (GC) Max. 0.1 %
2-Butanol (GC) Max. 0.1 %
Diethyl ether (GC) Max. 0.1 %
Diisopropyl alcohol Max. 0.1 %
1-propanol (GC) Max. 0.1 %
Impurities (GC) total Max. 1.0 %
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR