Heptane (mixture of isomers) 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

H2198-2.5L
BJH2198-2.5LEA 274.42 USD
BJH2198-2.5L
Heptane (mixture of isomers) 99%, Reagent Grade
Heptane (mixture of isomers)

  • ≥95% for general laboratory use.


Beilstein Number: 1730763

Formula: H₃C(CH₂)₅CH₃
Boiling Pt: 6.7 %(V)
Melting Pt: –90.5 °C
Density: 0.69 g/cm³ (15 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=d30a8083280d1915a43f51d93c49f9b76eabd579&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99 %
Refractive index (n 20/D) 1.386 - 1.388
IR spectrum complying
Appearance complying
Vapor Pressure 83 mmHg ( 37.7 °C)
Vapor Density 3.5 (vs air)
Explosion Limit 7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR