Methanol ≥99.9%, GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

GC230-4
BJGC230-4CS 1225 USD
BJGC230-4
Methanol ≥99.9%, GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Methanol

For trace analysis at or below the part per billion level. Ideal for EPA protocols.


  • Less than 1 ppb trace contamination for
  • Semi-volatiles with a molecular weight
  • Greater than toluene (92)
  • Ideal for EPA protocols
  • Actual lot chromatograms available upon request

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC (as lindane) 2 ppt max.
Flame Ionization GC (as nonane) 1 ppb max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3280 - 1.3288
Residue 1 mg/L max.
Water by Karl Fischer Titration 0.1 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR