Acetone ≥99.9% (dried basis), GC2® for gas chromatography, for pesticide residue analysis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

GC010-4
BJGC010-4CS 1220 USD
BJGC010-4
Acetone ≥99.9% (dried basis), GC2® for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Acetone

B&J GC2 Solvents are developed for trace analysis at or below the part-per-billion level, using capillary gas chromatography. Each lot is performance tested to meet demanding GC specifications.

Samples of every B&J GC2 solvent are concentrated 1000-fold and tested by temperature-programmed capillary GC-FID. This analysis detects both low-boiling and high-boiling molecules, anything eluting from toluene through diesel fuel and other semi-volatile compounds of environmental interest.

Features:
Less than 1 ppb trace contamination for semi-volatiles with a molecular weight greater than toluene (92).
Ideal for EPA protocols
Actual lot chromatograms available upon request

Electron capture GC: No residue peaks greater than two ng/L as lindane.

Flame ionization GC: No peaks greater than one ng/mL, having a retention time of greater than diacetone alcohol, on a crossbonded 5% diphenyl/95% dimethyl polysiloxane capillary column for a splitless injection of a 1000-fold sample concentrate after exchange into hexane.


  • Less than 1 ppb trace contamination for
  • Semi-volatiles with a molecular weight
  • Greater than toluene (92)
  • Ideal for EPA protocols
  • Actual lot chromatograms available upon request

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.79 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC (as lindane) 2 ppt max.
Flame Ionization GC (as nonane) 1 ppb max.
GC Analysis (excluding water) 99.9 % min.
Refractive Index (20ºC) 1.3583 - 1.3589
Residue 1 mg/L max.
Water by Karl Fischer Titration 0.5 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR