Glycerine ≥99.5% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

G7893-4X4L
BJG7893-4LCS 2134 USD
BJG7893-4L
Glycerine ≥99.5% ACS, Riedel-de Haën™
Glycerine

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: HOCH₂CH(OH)CH₂OH
Boiling Pt: 290 °C (1.013 hPa)
Melting Pt: 18 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ignition Residue ≤0.0050%
Identity (IR) complying
Assay (GC) Min. 99.50 %
Residue on ignition Max. 0.005 %
Water (Karl Fischer) Max. 0.5 %
Indifference complying
Heavy metals (as Pb) Max. 2 ppm
Sulfate (SO₄) Max. 0.001 %
Halogene compounds (as Cl) Max. 0.003 %
Acrolein and glucose complying
Fatty acids and ester (as butyric acid) Max. 0.05 %
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR