n-Dodecane ≥99%, Reagent Grade
Supplier: Honeywell Research Chemicals
D221104-500ML
BJD221104100MLEA
122.48
USD
BJD221104100ML
BJD221104500ML
n-Dodecane ≥99%, Reagent Grade
n-Dodecane
Specification Test Results
Vapor Pressure | 1 mmHg ( 47.8 °C) |
IR spectrum | complying |
Assay (GC) | Min. 99 % |
Refractive index (n 20/D) | 1.420 - 1.424 |
APHA | Max. 20 |
Vapor Density | 5.96 (vs air) |
Autoignition Temperature | 401 °F |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...