Toluene ≥99.8%, B&J Brand™ for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

CS347-200 CS347-56
BJCS347-200EA 5417.86 USD
BJCS347-200 BJCS347-56
Toluene ≥99.8%, B&J Brand™ for HPLC, for spectrophotometry, Burdick & Jackson™
Toluene

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3 II
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Benzene 500 mg/L max.
Water by Karl Fischer Titration 0.01 % max.
GC Analysis 99.8 % min.
Refractive Index (20ºC) 1.4963 - 1.4971
Residue 1 mg/L max.
UV Absorbance @ 300 nm 0.12 AU max.
UV Absorbance @ 325 nm 0.025 AU max.
UV Absorbance @ 350 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 284 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR