Methanol ≥99.9% ACS for HPLC

Supplier: Honeywell Research Chemicals
Danger

CS230AH200
BJCS230AH200EA 2303.83 USD
BJCS230AH200
Methanol ≥99.9% ACS for HPLC
Methanol
Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm3
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR