Hexane (mixture of isomers) ≥60,0% (n-hexane) ≥99.9%, B&J Brand™ for gas chromatography, not for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hexanes, Isohexane

CS211-56 CS211-200
BJCS211-56EA 1792.91 USD
BJCS211-56 BJCS211-200
Hexane (mixture of isomers) ≥60,0% (n-hexane) ≥99.9%, B&J Brand™ for gas chromatography, not for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™
Hexane (mixture of isomers)

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 7.7 %(V)
Melting Pt: -95
Density: 0.659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis (n-Hexane) 60 % min.
GC Analysis (total C6 isomers) 99.9 % min.
Residue 1 mg/L max.
Water by Karl Fisher Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR