Heptane (mixture of isomers) ≥99.0%, B&J Brand™ for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

CS207-56
BJCS207-56EA 4580.87 USD
BJCS207-56
Heptane (mixture of isomers) ≥99.0%, B&J Brand™ for HPLC, for spectrophotometry, Burdick & Jackson™
Heptane (mixture of isomers)

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 98.4 °C
Melting Pt: –90.6 °C
Density: 0.684 g/cm3
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=d30a8083280d1915a43f51d93c49f9b76eabd579&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis (n-Heptane) 99 % min.
Water by Karl Fischer Titration 0.01 % max.
GC Analysis (toal C7 isomers) 99.9 % min.
Refractive Index (20ºC) 1.3872 - 1.3884
Residue 1 mg/L max.
UV Absorbance @ 225 nm 0.1 AU max.
UV Absorbance @ 250 nm 0.01 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 200 nm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR