Acetone ≥99.9% (by GC, corrected for water content) for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

CS011-200
BJCS011-200EA 4912.02 USD
BJCS011-200
Acetone ≥99.9% (by GC, corrected for water content) for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Acetone

Acetone is a high purity solvent for pesticide residue analysis in pressure dispense system.


Packaging: Packed in a stainless steel pressure dispense system (SSPDS).

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56 °C
Melting Pt: –94.8 °C
Density: 0.79 g/cm3
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
Water by Karl Fischer Titration 0.5 % max.
GC Analysis (excluding water) 99.9 % min.
Refractive Index (20ºC) 1.3583 - 1.3589
Residue 1 mg/L max.
UV Absorbance @ 340 nm 0.06 AU max.
UV Absorbance @ 350 nm 0.01 AU max.
UV Absorbance @ 375 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 330 nm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR