Calcium chloride, anhydrous ≥97%, powder, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Chloro calcium, Calcium dichloride, CaCl2
C4901-1KG
C4901-100G
C4901-2.5KG
C4901-500G
BJC4901-1KGEA
322.3
USD
BJC4901-1KG
BJC4901-100G
BJC4901-2.5KG
BJC4901-500G
BJC4901-5KG
Calcium chloride, anhydrous ≥97%, powder, Fluka™
Calcium chloride
Formula:
CaCl₂ Boiling Pt: 1.900 °C (1.013 hPa) Melting Pt: 765 °C Density: 2.150 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 |
Specification Test Results
Colour of the solution | complying |
Clearness of the solution | complying |
Assay | Min. 97.0 % |
Mg and alkali salts | Max. 3.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...