5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
5-Ethyltio-1H-Tetrazole
BR725-4X2.5GL
BR725-20
BJBR725-1CS
12191.5
USD
BJBR725-1
BJBR725-2.5GL
BJBR725-20
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazole (ETT)
Reagents are specifically processed and purified to ensure the high synthesis efficiency
- Ideal for oligonucleotide synthesis
- Extremely low water content
- High purity reagents
- Low amine specifications
Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.
Formula:
C₃H₆N₄S MW: 130.17 g/mol Boiling Pt: 82 °C (main component) Melting Pt: –46 °C (main component) Density: 0.809 g/cm³ (20 °C) Flash Pt: 13 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068733 CAS Number: 89797-68-2 UN: 1648 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...