Dichloroacetic acid (DCA) 10% (v/v) in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
DCA Deblock, DCA
BR676ADAK-4
BR676-4X2.5L
BJBR676ADAK-4CS
1383.31
USD
BJBR676ADAK-4
BJBR676-4X2.5L
Dichloroacetic acid (DCA) 10% (v/v) in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Dichloroacetic acid (DCA)
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Formula:
C₂H₂Cl₂O₂ MW: 128.94 g/mol Boiling Pt: 194 Melting Pt: -4 Density: 0.9356 g/cm3 (20 °C) Flash Pt: 8.33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004223 CAS Number: 79-43-6 UN: 2924 ADR: 8 II Merck Index: 13,03075 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...