Oxidation reagent 0.025 M I₂ in 45/45/10 Pyridine/Acetonitrile/Water (v/v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

BR673AK-4
BJBR673AK-4CS 1484.03 USD
BJBR673AK-4
Oxidation reagent 0.025 M I₂ in 45/45/10 Pyridine/Acetonitrile/Water (v/v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Iodine

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 80.56 °C
Density: 0.8977 g/cm3 (25 °C)
Flash Pt: 8.89 °C
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR