Capping Reagent 15% N-Methylimidazole in Acetonitrile, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
BR660-4
BJBR660-4CS
6370.3
USD
BJBR660-4
Capping Reagent 15% N-Methylimidazole in Acetonitrile, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Boiling Pt:
67.2 °C Melting Pt: –43.8 °C (main component) Density: 0.8159 g/cm3 (25 °C) Flash Pt: 10.56 °C Storage Temperature: Ambient |
UN:
2924 ADR: 3 II |