Capping Reagent 50/30/20 Acetonitrile/Pyridine/NMI (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
BR656-4
BJBR656-4CS
4052.44
USD
BJBR656-4
Capping Reagent 50/30/20 Acetonitrile/Pyridine/NMI (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Reagents are specifically processed and purified to ensure the high synthesis efficiency
- Ideal for oligonucleotide synthesis
- Extremely low water content
- High purity reagents
- Low amine specifications
Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.
Density:
0.8936 g/cm3 Flash Pt: 4.4 °C |
UN:
2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...