Acetic anhydride ≥99.5%, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
BR649-4
BJBR649-4CS
1685
USD
BJBR649-4
Acetic anhydride ≥99.5%, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Acetic anhydride
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Beilstein Number: 385737
Formula:
(CH₃CO)₂O Boiling Pt: 140 °C Melting Pt: –73.1 °C Density: 1.083 g/cm³ (20 °C) Flash Pt: 10.3 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00008705 CAS Number: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...