Tetrahydrofuran ≥99.9% stabilized, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

BB343-4 BN343-20
BJBB343-4CS 2066.97 USD
BJBB343-4 BJBN343-20
Tetrahydrofuran ≥99.9% stabilized, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Tetrahydrofuran

BioSyn« products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.


  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 11.8 %(V)
Melting Pt: –108.5 °C
Density: 0.8892 g/cm³ (20 °C)
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Peroxides (as H₂O₂) at time of packaging 2 mg/L max.
Refractive Index (20ºC) 1.4065 - 1.4075
Water by Karl Fischer Titration 100 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR