5-(Ethylthio)-1H-tetrazole (ETT) 0.25M in acetonitrile, BioSyn™ for biochemistry, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 5-Ethyltio-1H-Tetrazole

BR721-4 SR721-1 SR721-1 BC721-200 BC721-56 BR721-20 BR721-4X2.5L SR721-2.5
BJBR721-2CS 60705.6 USD
BJBR721-2 BJBR721-4 BASR721-1NAN BJSR721-1 BJBC721-200 BJBC721-56 BJBR721-20 BJBR721-4X2.5L BJSR721-2.5
5-(Ethylthio)-1H-tetrazole (ETT) 0.25M in acetonitrile, BioSyn™ for biochemistry, for DNA/RNA synthesis, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazole (ETT)

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Formula: C₃H₆N₄S
MW: 130.17 g/mol
Boiling Pt: 82 °C
Density: 0.790 g/cm3
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00068733
CAS Number: 89797-68-2
UN: 1648
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR